Product Details

SNP ID
rs59334819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85174329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACGGACTGAGAAGAGATCTGCTC[A/C]GGGCCCAGAGGTGGCAGCAATCCAG
Phenotype
MIM: 616886
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSE1 PubMed Links

Gene Details

Gene
GSE1
Gene Name
Gse1 coiled-coil protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134473.2 Intron NP_001127945.1
NM_001278184.1 Intron NP_001265113.1
NM_014615.3 Intron NP_055430.1
XM_005255859.4 Intron XP_005255916.3
XM_005255860.3 Intron XP_005255917.3
XM_005255861.4 Intron XP_005255918.3
XM_005255863.4 Intron XP_005255920.1
XM_005255864.4 Intron XP_005255921.1
XM_005255865.4 Intron XP_005255922.1
XM_011522965.2 Intron XP_011521267.1
XM_011522966.2 Intron XP_011521268.1
XM_017023083.1 Intron XP_016878572.1
XM_017023084.1 Intron XP_016878573.1

View Full Product Details