Product Details

SNP ID
rs61758746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82658086 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGAGCTTGTCCACCAGGTGCACC[C/G]GCGTGCAGGACACGACCGCCCGGCA
Phenotype
MIM: 609226
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB40B PubMed Links

Gene Details

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 852 Missense Mutation CCG,CGG P205R NP_006813.1
XM_005256334.4 852 Missense Mutation CCG,CGG P145R XP_005256391.1
XM_005256335.3 852 Intron XP_005256392.1
XM_006722271.3 852 Missense Mutation CCG,CGG P165R XP_006722334.1
XM_011523528.2 852 Missense Mutation CCG,CGG P188R XP_011521830.1
XM_017024042.1 852 Intron XP_016879531.1
XM_017024043.1 852 Missense Mutation CCG,CGG P145R XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details