Product Details

SNP ID
rs59654138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35306446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATCCCCAAGTTTCTGTCTTGAG[A/G]ATGACAGAGCTTTGTCTAGAAAGGG
Phenotype
MIM: 604700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOM1 PubMed Links

Gene Details

Gene
TOM1
Gene Name
target of myb1 membrane trafficking protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135729.1 Intron NP_001129201.1
NM_001135730.1 Intron NP_001129202.1
NM_001135732.1 Intron NP_001129204.1
NM_005488.2 Intron NP_005479.1
XM_011529818.2 Intron XP_011528120.1
XM_011529820.2 Intron XP_011528122.1
XM_017028529.1 Intron XP_016884018.1
XM_017028530.1 Intron XP_016884019.1

View Full Product Details