Product Details
- SNP ID
-
rs56384521
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:109913453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCTCGCAGGAAATGGGAATTCT[A/G]CTTGTGTGGGTAAGTACTCCAATGA
- Phenotype
-
MIM: 131530
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EGF
PubMed Links
Gene Details
- Gene
- EGF
- Gene Name
- epidermal growth factor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001178130.2 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
NP_001171601.1 |
NM_001178131.2 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
NP_001171602.1 |
NM_001963.5 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
NP_001954.2 |
XM_005262796.2 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_005262853.1 |
XM_011531707.1 |
571 |
UTR 5 |
|
|
XP_011530009.1 |
XM_017007845.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863334.1 |
XM_017007846.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863335.1 |
XM_017007847.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863336.1 |
XM_017007848.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863337.1 |
XM_017007849.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863338.1 |
XM_017007850.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863339.1 |
XM_017007851.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863340.1 |
XM_017007852.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863341.1 |
XM_017007853.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863342.1 |
XM_017007854.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863343.1 |
XM_017007855.1 |
571 |
Missense Mutation |
ACT,GCT |
T40A |
XP_016863344.1 |
View Full Product Details