Product Details

SNP ID
rs62246207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46712131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGAAGGAGGCATGGAAAACAGTA[A/G]TGTTTAATTGAGCACCTCATCTCCA
Phenotype
MIM: 607950 MIM: 607237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS46 PubMed Links

Gene Details

Gene
PRSS46
Gene Name
protease, serine 46
There are no transcripts associated with this gene.

Gene
PRSS50
Gene Name
protease, serine 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013270.4 1332 UTR 3 NP_037402.1
Gene
TMIE
Gene Name
transmembrane inner ear
There are no transcripts associated with this gene.

View Full Product Details