Product Details

SNP ID
rs57859638
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51896834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGTGGTTTGGGAGGTCATACTT[C/T]TTCTGATTTCTCATTGGCATTGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQCF1 PubMed Links

Gene Details

Gene
IQCF1
Gene Name
IQ motif containing F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152397.2 267 Missense Mutation NP_689610.2
XM_011533366.2 267 Missense Mutation XP_011531668.1
XM_017005729.1 267 Missense Mutation XP_016861218.1

View Full Product Details