Product Details

SNP ID
rs58207137
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:66813691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTATTTTCATCTACTAAAAACTA[C/T]GTAATTATAAACTCTGGAAAACAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDR78 PubMed Links

Gene Details

Gene
WDR78
Gene Name
WD repeat domain 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024763.4 2800 UTR 3 NP_079039.4
NM_207014.2 2800 Intron NP_996897.2
XM_005271212.3 2800 UTR 3 XP_005271269.1
XM_011542161.2 2800 UTR 3 XP_011540463.1
XM_011542162.2 2800 Intron XP_011540464.1
XM_017002353.1 2800 UTR 3 XP_016857842.1
XM_017002354.1 2800 UTR 3 XP_016857843.1
XM_017002355.1 2800 UTR 3 XP_016857844.1
XM_017002356.1 2800 UTR 3 XP_016857845.1
XM_017002357.1 2800 Intron XP_016857846.1

View Full Product Details