Product Details

SNP ID
rs55766259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65654742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCTCTGACAGCGTTCCTTCCCCA[C/G]CCTGGGTGGGCTTGGGGGCAGGTGG
Phenotype
MIM: 164014 MIM: 602180
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4489 PubMed Links

Gene Details

Gene
MIR4489
Gene Name
microRNA 4489
There are no transcripts associated with this gene.

Gene
RELA
Gene Name
RELA proto-oncogene, NF-kB subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145138.1 1200 Missense Mutation GCT,GGT A428G NP_001138610.1
NM_001243984.1 1200 Missense Mutation GCT,GGT A362G NP_001230913.1
NM_001243985.1 1200 Intron NP_001230914.1
NM_021975.3 1200 Missense Mutation GCT,GGT A431G NP_068810.3
XM_011545206.1 1200 Missense Mutation GCT,GGT A360G XP_011543508.1
XM_011545207.1 1200 Missense Mutation GCT,GGT A325G XP_011543509.1
Gene
SIPA1
Gene Name
signal-induced proliferation-associated 1
There are no transcripts associated with this gene.

View Full Product Details