Product Details

SNP ID
rs72754652
Assay Type
Functionally Tested
NCBI dbSNP Submissions
24
Location
Chr.1:239397912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTAACTGGGGGCACACAATAAA[C/G]CATTTGTTGAAAAGAAAATTCTTTT
Phenotype
MIM: 118494
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CHRM3 PubMed Links

Gene Details

Gene
CHRM3
Gene Name
cholinergic receptor muscarinic 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000740.2 Intron NP_000731.1
XM_005273032.3 Intron XP_005273089.1
XM_011544041.2 Intron XP_011542343.1
XM_011544043.2 Intron XP_011542345.1
XM_011544044.2 Intron XP_011542346.1
XM_011544045.2 Intron XP_011542347.1
XM_011544046.2 Intron XP_011542348.1
XM_011544047.2 Intron XP_011542349.1
XM_017000152.1 Intron XP_016855641.1
XM_017000153.1 Intron XP_016855642.1
XM_017000154.1 Intron XP_016855643.1
XM_017000155.1 Intron XP_016855644.1
XM_017000156.1 Intron XP_016855645.1
XM_017000157.1 Intron XP_016855646.1
XM_017000158.1 Intron XP_016855647.1
XM_017000159.1 Intron XP_016855648.1
XM_017000160.1 Intron XP_016855649.1
XM_017000161.1 Intron XP_016855650.1
XM_017000162.1 Intron XP_016855651.1
XM_017000163.1 Intron XP_016855652.1

View Full Product Details