Product Details

SNP ID
rs73267929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:1045641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGAGGCACTGTTAATAATTAGAC[A/G]AAGTGCACACTCACAGTACCCATGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf50 PubMed Links

Gene Details

Gene
C7orf50
Gene Name
chromosome 7 open reading frame 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134395.1 Intron NP_001127867.1
NM_001134396.1 Intron NP_001127868.1
NM_001318252.1 Intron NP_001305181.1
NM_032350.5 Intron NP_115726.1
XM_006715793.2 Intron XP_006715856.1
XM_011515580.2 Intron XP_011513882.1
XM_011515581.2 Intron XP_011513883.1
XM_011515582.2 Intron XP_011513884.1
XM_011515583.2 Intron XP_011513885.1
XM_011515584.2 Intron XP_011513886.1
XM_017012719.1 Intron XP_016868208.1
XM_017012720.1 Intron XP_016868209.1
XM_017012721.1 Intron XP_016868210.1
XM_017012722.1 Intron XP_016868211.1
Gene
GPR146
Gene Name
G protein-coupled receptor 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303473.1 Intron NP_001290402.1
NM_001303474.1 Intron NP_001290403.1
NM_138445.3 Intron NP_612454.1
XM_005249613.2 Intron XP_005249670.1
XM_006715642.2 Intron XP_006715705.1
XM_006715643.2 Intron XP_006715706.1
XM_011515116.2 Intron XP_011513418.1

View Full Product Details