Product Details

SNP ID
rs68102548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45935173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGGGGGGTTAGGCAGTCCCCCC[C/T]CAGTTATCTCTGGGAGACAAGTGTC
Phenotype
MIM: 601991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOVA2 PubMed Links

Gene Details

Gene
NOVA2
Gene Name
NOVA alternative splicing regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002516.3 6217 UTR 3 NP_002507.1
XM_006723230.3 6217 Intron XP_006723293.1
XM_017026838.1 6217 Intron XP_016882327.1
XM_017026839.1 6217 Intron XP_016882328.1
XM_017026840.1 6217 Intron XP_016882329.1

View Full Product Details