Product Details

SNP ID
rs73546793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21508306 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCCTGACCCCGGTGAGATGAGG[C/T]GAGAAATTGCAAAGTAAAATGCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00664 PubMed Links

Gene Details

Gene
LINC00664
Gene Name
long intergenic non-protein coding RNA 664
There are no transcripts associated with this gene.

Gene
ZNF429
Gene Name
zinc finger protein 429
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001415.2 Intron NP_001001415.2
XM_017026748.1 Intron XP_016882237.1
XM_017026749.1 Intron XP_016882238.1
XM_017026750.1 Intron XP_016882239.1
XM_017026751.1 Intron XP_016882240.1
XM_017026752.1 Intron XP_016882241.1
XM_017026753.1 Intron XP_016882242.1
XM_017026754.1 Intron XP_016882243.1
XM_017026755.1 Intron XP_016882244.1
XM_017026756.1 Intron XP_016882245.1
XM_017026757.1 Intron XP_016882246.1

View Full Product Details