Product Details

SNP ID
rs71439225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38666317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCAAAGACAGGCAGGGGAGAGC[C/T]TCGGACGTGGTGCTTGGCTTCGCCG
Phenotype
MIM: 137030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALM PubMed Links

Gene Details

Gene
GALM
Gene Name
galactose mutarotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138801.2 408 Silent Mutation GCC,GCT A52A NP_620156.1
XM_011532540.1 408 Silent Mutation GCC,GCT A52A XP_011530842.1

View Full Product Details