Product Details

SNP ID
rs73011419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174299153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGTGACTTAAAAAGAAGCTTAC[A/G]GAACCTAGAACAGGTGCTCCGCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP44 PubMed Links

Gene Details

Gene
CEP44
Gene Name
centrosomal protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040157.2 370 Missense Mutation CAG,CGG Q11R NP_001035247.1
NM_001145314.1 370 Missense Mutation CAG,CGG Q11R NP_001138786.1
XM_011532285.2 370 Missense Mutation CAG,CGG Q11R XP_011530587.1
XM_011532286.2 370 Missense Mutation CAG,CGG Q11R XP_011530588.1
XM_011532287.2 370 Missense Mutation CAG,CGG Q11R XP_011530589.1
XM_017008654.1 370 Missense Mutation CAG,CGG Q11R XP_016864143.1
XM_017008655.1 370 Missense Mutation CAG,CGG Q11R XP_016864144.1
XM_017008656.1 370 Missense Mutation CAG,CGG Q11R XP_016864145.1
XM_017008657.1 370 Missense Mutation CAG,CGG Q11R XP_016864146.1
XM_017008658.1 370 UTR 5 XP_016864147.1
XM_017008659.1 370 UTR 5 XP_016864148.1
XM_017008660.1 370 UTR 5 XP_016864149.1

View Full Product Details