Product Details

SNP ID
rs73269515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54578119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACCTCATTTATTCTTCTCAATA[A/G]CCCTATGAGATGAGGTAGGAACTGC
Phenotype
MIM: 610747
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAMD4A PubMed Links

Gene Details

Gene
SAMD4A
Gene Name
sterile alpha motif domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161576.2 Intron NP_001155048.2
NM_001161577.1 Intron NP_001155049.1
NM_015589.5 Intron NP_056404.4

View Full Product Details