Product Details

SNP ID
rs73200921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:17248031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGTTCTGTTGTCTTCCTGTGAG[C/T]TTTTCATTTTATCATGGTCAGTTTA
Phenotype
MIM: 604913 MIM: 609927
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNOT7 PubMed Links

Gene Details

Gene
CNOT7
Gene Name
CCR4-NOT transcription complex subunit 7
There are no transcripts associated with this gene.

Gene
VPS37A
Gene Name
VPS37A, ESCRT-I subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145152.1 Intron NP_001138624.1
NM_152415.2 Intron NP_689628.2
XM_005273400.2 Intron XP_005273457.1
XM_005273401.2 Intron XP_005273458.1
XM_006716286.1 Intron XP_006716349.1
XM_017013021.1 Intron XP_016868510.1
XM_017013022.1 Intron XP_016868511.1
XM_017013023.1 Intron XP_016868512.1
XM_017013024.1 Intron XP_016868513.1
XM_017013025.1 Intron XP_016868514.1
XM_017013026.1 Intron XP_016868515.1

View Full Product Details