Product Details

SNP ID
rs72657942
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.1:34863757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAAGCCCATTTCCTCATCTGT[A/G]AATGGGGGTGAAAACAATATCTGTT
Phenotype
MIM: 611413
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DLGAP3 PubMed Links

Gene Details

Gene
DLGAP3
Gene Name
DLG associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080418.2 5244 Intron NP_001073887.1
XM_011541879.2 5244 UTR 3 XP_011540181.1
XM_011541880.2 5244 UTR 3 XP_011540182.1
Gene
SMIM12
Gene Name
small integral membrane protein 12
There are no transcripts associated with this gene.

View Full Product Details