Product Details

SNP ID
rs73543099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73950863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCAATTTCCGAGTTGAGTGAC[C/T]TTTCACAGGGTCGCAGAATCAGCCC
Phenotype
MIM: 610263
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 243 UTR 5 NP_705842.2
XM_005273984.3 243 UTR 5 XP_005274041.1
XM_011545004.2 243 UTR 5 XP_011543306.1
XM_011545005.2 243 UTR 5 XP_011543307.1
XM_011545007.2 243 Intron XP_011543309.1
XM_011545009.2 243 UTR 5 XP_011543311.1
XM_011545013.2 243 UTR 5 XP_011543315.1
XM_011545014.2 243 UTR 5 XP_011543316.1
XM_011545015.2 243 Intron XP_011543317.1
XM_017017675.1 243 Intron XP_016873164.1
XM_017017676.1 243 Intron XP_016873165.1
XM_017017677.1 243 Intron XP_016873166.1
XM_017017678.1 243 Intron XP_016873167.1
XM_017017679.1 243 UTR 5 XP_016873168.1

View Full Product Details