Product Details

SNP ID
rs77464454
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9960999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTAGGAAAATAAAAATTGTCAA[A/G]GAACTACATCAGCATCGCGTTTGAA
Phenotype
MIM: 609786
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRHL1 PubMed Links

Gene Details

Gene
GRHL1
Gene Name
grainyhead like transcription factor 1
There are no transcripts associated with this gene.

View Full Product Details