Product Details

SNP ID
rs79052831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:169128119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTACAGTCAAGTCCAGATGTCT[A/G]TACAACACATAGTGGCCCCTATCAA
Phenotype
MIM: 600073
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRP2 PubMed Links

Gene Details

Gene
LRP2
Gene Name
LDL receptor related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004525.2 14597 UTR 3 NP_004516.2
XM_011511183.2 14597 UTR 3 XP_011509485.1
XM_011511184.2 14597 UTR 3 XP_011509486.1

View Full Product Details