Product Details

SNP ID
rs4520
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:116830819 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCTCCCTTCTCAGCTTCATGCAGGG[C/T]TACATGAAGCACGCCACCAAGACCG
Phenotype
MIM: 107680 MIM: 107690 MIM: 107720
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APOA1 PubMed Links

Gene Details

Gene
APOA1
Gene Name
apolipoprotein A1
There are no transcripts associated with this gene.

Gene
APOA1-AS
Gene Name
APOA1 antisense RNA
There are no transcripts associated with this gene.

Gene
APOA4
Gene Name
apolipoprotein A4
There are no transcripts associated with this gene.

Gene
APOC3
Gene Name
apolipoprotein C3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000040.1 148 Silent Mutation GGC,GGT G34G NP_000031.1

View Full Product Details