Product Details
- SNP ID
-
rs7197178
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:55728953 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTAATTGTGTTGGCAAAGGGTGATG[A/T]CCCAATGTGTCAGCATTTGCCAAAT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CES1P2
PubMed Links
Gene Details
- Gene
- CES1P2
- Gene Name
- carboxylesterase 1 pseudogene 2
There are no transcripts associated with this gene.
View Full Product Details