Product Details

SNP ID
rs61734600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67405440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGGCATCCCGTCTGCCCTGCG[C/T]GCCCGATGCTGGCCCCTGTTGTGTG
Phenotype
MIM: 176875 MIM: 603761 MIM: 610831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1CA PubMed Links

Gene Details

Gene
PPP1CA
Gene Name
protein phosphatase 1 catalytic subunit alpha
There are no transcripts associated with this gene.

Gene
RAD9A
Gene Name
RAD9 checkpoint clamp component A
There are no transcripts associated with this gene.

Gene
TBC1D10C
Gene Name
TBC1 domain family member 10C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256508.1 610 Silent Mutation CGC,CGT R98R NP_001243437.1
NM_198517.3 610 Silent Mutation CGC,CGT R98R NP_940919.1
XM_006718538.3 610 Silent Mutation CGC,CGT R98R XP_006718601.1
XM_006718539.3 610 Silent Mutation CGC,CGT R98R XP_006718602.1
XM_006718540.3 610 Silent Mutation CGC,CGT R98R XP_006718603.1
XM_006718541.3 610 UTR 5 XP_006718604.1
XM_006718542.3 610 UTR 5 XP_006718605.1
XM_006718543.2 610 UTR 5 XP_006718606.1
XM_006718544.3 610 UTR 5 XP_006718607.1
XM_011545001.2 610 UTR 5 XP_011543303.1
XM_011545002.2 610 Silent Mutation CGC,CGT R98R XP_011543304.1

View Full Product Details