Product Details

SNP ID
rs35311561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103721790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTATTCATTTTCCTCACTAAAC[A/G]TCTTGAGGCCTGTTTCATTGGGTTG
Phenotype
MIM: 600675 MIM: 613504
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107984648 PubMed Links

Gene Details

Gene
LOC107984648
Gene Name
uncharacterized LOC107984648
There are no transcripts associated with this gene.

Gene
XRCC3
Gene Name
X-ray repair cross complementing 3
There are no transcripts associated with this gene.

Gene
ZFYVE21
Gene Name
zinc finger FYVE-type containing 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198953.1 Intron NP_001185882.1
NM_024071.3 Intron NP_076976.1

View Full Product Details