Product Details

SNP ID
rs3765654
Assay Type
Validated
NCBI dbSNP Submissions
35
Location
Chr.1:74244027 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CAAAAGAAGAGAAAATGGAATTTCT[C/G]TCCTAGGAATGATTGCTTAAATATC
Phenotype
MIM: 613932
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FPGT-TNNI3K PubMed Links

Gene Details

Gene
FPGT-TNNI3K
Gene Name
FPGT-TNNI3K readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112808.2 Intron NP_001106279.2
NM_001199327.1 Intron NP_001186256.2
Gene
TNNI3K
Gene Name
TNNI3 interacting kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015978.2 Intron NP_057062.1

View Full Product Details