Product Details

SNP ID
rs538808
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:11542403 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGTTATTCATCCAGGGTTAGCTCCA[A/G]TATGTCTTTCAATATGATCTTAATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM170B PubMed Links

Gene Details

Gene
TMEM170B
Gene Name
transmembrane protein 170B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100829.2 Intron NP_001094299.1

View Full Product Details