Product Details

SNP ID
rs1566823
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:127260616 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATCTCAAAAGGCAGCCTGGATTGC[A/G]GTCTTTTGCAATCAAGAACCTGGGA
Phenotype
MIM: 133510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERCC3 PubMed Links

Gene Details

Gene
ERCC3
Gene Name
ERCC excision repair 3, TFIIH core complex helicase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000122.1 Intron NP_000113.1
NM_001303416.1 Intron NP_001290345.1
NM_001303418.1 Intron NP_001290347.1
XM_011510794.1 Intron XP_011509096.1
XM_011510795.1 Intron XP_011509097.1
XM_017003583.1 Intron XP_016859072.1

View Full Product Details