Product Details

SNP ID
rs2659005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81244914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCAACCATGAGGAGTGCAAAGGG[C/T]GGCACCGGCCAGTGCCCCTGGACAC
Phenotype
MIM: 616525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf89 PubMed Links

Gene Details

Gene
C17orf89
Gene Name
chromosome 17 open reading frame 89
There are no transcripts associated with this gene.

Gene
SLC38A10
Gene Name
solute carrier family 38 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037984.2 4420 UTR 3 NP_001033073.1
NM_138570.3 4420 Intron NP_612637.1
XM_005257019.1 4420 Intron XP_005257076.1
XM_011524288.1 4420 Intron XP_011522590.1
XM_011524289.1 4420 Intron XP_011522591.1
XM_011524290.1 4420 Intron XP_011522592.1
Gene
TEPSIN
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
There are no transcripts associated with this gene.

View Full Product Details