Product Details

SNP ID
rs11177615
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:69360409 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGTGTGAGGATAACAATAACTGTA[A/G]CAAACAGTCGCTGAGCGCTTACCCG
Phenotype
MIM: 153450 MIM: 602116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYZ PubMed Links

Gene Details

Gene
LYZ
Gene Name
lysozyme
There are no transcripts associated with this gene.

Gene
YEATS4
Gene Name
YEATS domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300950.1 Intron NP_001287879.1
NM_006530.3 Intron NP_006521.1

View Full Product Details