Product Details

SNP ID
rs3860064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37899532 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCATCTCCTGTGATAAGTGACTG[A/G]AGCTCTCCGGGCTTCAGTTTCCTAG
Phenotype
MIM: 609850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBC1D1 PubMed Links

Gene Details

Gene
TBC1D1
Gene Name
TBC1 domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253912.1 Intron NP_001240841.1
NM_001253913.1 Intron NP_001240842.1
NM_001253914.1 Intron NP_001240843.1
NM_001253915.1 Intron NP_001240844.1
NM_015173.3 Intron NP_055988.2
XM_005262646.2 Intron XP_005262703.1
XM_005262647.2 Intron XP_005262704.1
XM_005262649.2 Intron XP_005262706.1
XM_006714001.2 Intron XP_006714064.1
XM_011513659.2 Intron XP_011511961.1
XM_011513660.2 Intron XP_011511962.1
XM_011513662.2 Intron XP_011511964.1
XM_011513663.2 Intron XP_011511965.1
XM_011513664.2 Intron XP_011511966.1
XM_011513665.2 Intron XP_011511967.1
XM_011513666.2 Intron XP_011511968.1
XM_011513668.2 Intron XP_011511970.1
XM_011513670.2 Intron XP_011511972.1
XM_017007918.1 Intron XP_016863407.1
XM_017007919.1 Intron XP_016863408.1
XM_017007920.1 Intron XP_016863409.1
XM_017007921.1 Intron XP_016863410.1
XM_017007922.1 Intron XP_016863411.1

View Full Product Details