Product Details

SNP ID
rs35244470
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:651578 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GCCACTTGCTGCTGCCCTCCCCAGG[C/T]GTCACCCTCACCAGGCATCTGCACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371038 PubMed Links

Gene Details

Gene
LOC105371038
Gene Name
uncharacterized LOC105371038
There are no transcripts associated with this gene.

Gene
MCRIP2
Gene Name
MAPK regulated corepressor interacting protein 2
There are no transcripts associated with this gene.

Gene
WDR90
Gene Name
WD repeat domain 90
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145294.4 Intron NP_660337.3
XM_017023023.1 Intron XP_016878512.1
XM_017023024.1 Intron XP_016878513.1

View Full Product Details