Product Details

SNP ID
rs56344544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35068334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATAGAGCTCCACATCCCCACCAA[C/T]GAGGCAAGTCTCCCATGGAGGGAGC
Phenotype
MIM: 612019
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISX PubMed Links

Gene Details

Gene
ISX
Gene Name
intestine specific homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303508.1 Intron NP_001290437.1

View Full Product Details