Product Details

SNP ID
rs11964378
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:49500355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAGCAACCTCTCTGAACAAAAAT[C/T]CTAACATATGAAAAATAATTACTAG
Phenotype
MIM: 611506 MIM: 614763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPQ PubMed Links

Gene Details

Gene
CENPQ
Gene Name
centromere protein Q
There are no transcripts associated with this gene.

Gene
GLYATL3
Gene Name
glycine-N-acyltransferase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010904.1 Intron NP_001010904.1

View Full Product Details