Product Details

SNP ID
rs9367357
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:49511417 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCTCCAGTTTTTCCTCATAACGACC[C/T]GACCCACTCAAGTCTTTCTCTAGTC
Phenotype
MIM: 614763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLYATL3 PubMed Links

Gene Details

Gene
GLYATL3
Gene Name
glycine-N-acyltransferase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010904.1 Intron NP_001010904.1

View Full Product Details