Product Details

SNP ID
rs11995518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81526864 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAATCCACACACATTTTATATAA[C/T]GCTTACGATTATACATTTATCTCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FABP12 PubMed Links

Gene Details

Gene
FABP12
Gene Name
fatty acid binding protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105281.1 Intron NP_001098751.1
XM_005251285.4 Intron XP_005251342.1
XM_006716465.3 Intron XP_006716528.1
XM_011517577.2 Intron XP_011515879.1

View Full Product Details