Product Details

SNP ID
rs7552470
Assay Type
Validated
NCBI dbSNP Submissions
27
Location
Chr.1:179028648 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CTTCTGAATCTGACTCTTAGTCCTT[C/T]TGGGACTAAGTGCCAGGTGGTTTTG
Phenotype
MIM: 611063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM20B PubMed Links

Gene Details

Gene
FAM20B
Gene Name
family with sequence similarity 20 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324310.1 Intron NP_001311239.1
NM_001324311.1 Intron NP_001311240.1
NM_014864.3 Intron NP_055679.1
XM_017003001.1 Intron XP_016858490.1
XM_017003002.1 Intron XP_016858491.1

View Full Product Details