Product Details

SNP ID
rs11220207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125747298 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAGAGTGCAGGATGGACCCTGAA[A/G]GGCAGACTATAAGAACCCAATACAA
Phenotype
MIM: 606861
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PATE1 PubMed Links

Gene Details

Gene
PATE1
Gene Name
prostate and testis expressed 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138294.2 Intron NP_612151.1

View Full Product Details