Product Details

SNP ID
rs330543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110618923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAGGGAAGCTTTTCCGCTCTG[A/G]GGAGCAGATGCGGAGAGGACAAGCA
Phenotype
MIM: 615910
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAXD PubMed Links

Gene Details

Gene
NAXD
Gene Name
NAD(P)HX dehydratase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242881.1 Intron NP_001229810.1
NM_001242882.1 Intron NP_001229811.1
NM_001242883.1 Intron NP_001229812.1
NM_018210.3 Intron NP_060680.2

View Full Product Details