Product Details

SNP ID
rs11070266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40352575 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCTCAGACTCCAAGCTCCCACCC[A/C]CTTCCAATCCTGGTGGGAGGAGACA
Phenotype
MIM: 607503
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DISP2 PubMed Links

Gene Details

Gene
DISP2
Gene Name
dispatched RND transporter family member 2
There are no transcripts associated with this gene.

Gene
PHGR1
Gene Name
proline, histidine and glycine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145643.1 Intron NP_001139115.1

View Full Product Details