Product Details
- SNP ID
-
rs7122842
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:77134224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGATTACAGGTGTTAGCCACTGCA[C/T]CTGGCCTACAAATAACATCTTAAAT
- Phenotype
-
MIM: 602537
MIM: 276903
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CAPN5
PubMed Links
Gene Details
- Gene
- CAPN5
- Gene Name
- calpain 5
There are no transcripts associated with this gene.
- Gene
- MYO7A
- Gene Name
- myosin VIIA
View Full Product Details