Product Details

SNP ID
rs760982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46341517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGATATCCAAGGAACCTCTGA[C/T]CCTGTGCAGATCTGTGATATGTCTG
Phenotype
MIM: 607477 MIM: 610230
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTSE1 PubMed Links

Gene Details

Gene
GTSE1
Gene Name
G2 and S-phase expressed 1
There are no transcripts associated with this gene.

Gene
TRMU
Gene Name
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282782.1 Intron NP_001269711.1
NM_001282783.1 Intron NP_001269712.1
NM_001282784.1 Intron NP_001269713.1
NM_001282785.1 Intron NP_001269714.1
NM_018006.4 Intron NP_060476.2
XM_005261678.1 Intron XP_005261735.1
XM_005261681.1 Intron XP_005261738.1
XM_011530271.2 Intron XP_011528573.1
XM_011530272.1 Intron XP_011528574.1
XM_011530273.1 Intron XP_011528575.1
XM_011530274.2 Intron XP_011528576.1
XM_011530275.1 Intron XP_011528577.1

View Full Product Details