Product Details

SNP ID
rs445219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:47657495 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTTTTGAACACTGTATGACAATA[C/G]TTAAACTACAAAATTTTTTTTACAA
Phenotype
MIM: 601937 MIM: 610013
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NCOA3 PubMed Links

Gene Details

Gene
NCOA3
Gene Name
nuclear receptor coactivator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174087.1 4165 Intron NP_001167558.1
NM_001174088.1 4165 Intron NP_001167559.1
NM_006534.3 4165 Intron NP_006525.2
NM_181659.2 4165 Intron NP_858045.1
Gene
SULF2
Gene Name
sulfatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161841.1 4165 UTR 3 NP_001155313.1
NM_018837.3 4165 UTR 3 NP_061325.1
NM_198596.2 4165 UTR 3 NP_940998.2
XM_005260458.2 4165 UTR 3 XP_005260515.1
XM_006723827.1 4165 UTR 3 XP_006723890.1
XM_006723828.1 4165 UTR 3 XP_006723891.1
XM_006723829.2 4165 UTR 3 XP_006723892.1
XM_006723830.1 4165 UTR 3 XP_006723893.1
XM_011528913.1 4165 UTR 3 XP_011527215.1
XM_011528914.1 4165 UTR 3 XP_011527216.1
XM_011528915.1 4165 UTR 3 XP_011527217.1
XM_011528916.1 4165 UTR 3 XP_011527218.1
XM_017027955.1 4165 UTR 3 XP_016883444.1

View Full Product Details