Product Details

SNP ID
rs155250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2261159 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAAAAATTGTTTCAAAAACT[A/G]TAGTACACCCAAATACTGTTCTCAA
Phenotype
MIM: 600305 MIM: 606447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECI1 PubMed Links

Gene Details

Gene
ECI1
Gene Name
enoyl-CoA delta isomerase 1
There are no transcripts associated with this gene.

Gene
LOC106660606
Gene Name
uncharacterized LOC106660606
There are no transcripts associated with this gene.

Gene
MIR3677
Gene Name
microRNA 3677
There are no transcripts associated with this gene.

Gene
RNPS1
Gene Name
RNA binding protein with serine rich domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286625.1 Intron NP_001273554.1
NM_001286626.1 Intron NP_001273555.1
NM_001286627.1 Intron NP_001273556.1
NM_006711.4 Intron NP_006702.1
NM_080594.3 Intron NP_542161.1
XM_005255048.1 Intron XP_005255105.1
XM_005255049.3 Intron XP_005255106.1
XM_017022874.1 Intron XP_016878363.1

View Full Product Details