Product Details

SNP ID
rs12160956
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45347361 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATGTTAAAAATGCAGGGCCTTCAA[A/G]AGATTTTCTCAGATGAATACAATTT
Phenotype
MIM: 608685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM118A PubMed Links

Gene Details

Gene
FAM118A
Gene Name
family with sequence similarity 118 member A
There are no transcripts associated with this gene.

Gene
SMC1B
Gene Name
structural maintenance of chromosomes 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291501.1 Intron NP_001278430.1
NM_148674.4 Intron NP_683515.4
XM_011530144.2 Intron XP_011528446.1
XM_011530145.2 Intron XP_011528447.1

View Full Product Details