Product Details
- SNP ID
-
rs958075
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:25614487 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CCTAATAGTCTAGAGAAAATTCTCC[A/G]GTTCCAAGACGATGCCTTGCTATGT
- Phenotype
-
MIM: 610763
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NANP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147838290] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NANP
- Gene Name
- N-acetylneuraminic acid phosphatase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152667.2 |
2249 |
UTR 3 |
|
|
NP_689880.1 |
- Gene
- ZNF337-AS1
- Gene Name
- ZNF337 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details