Product Details

SNP ID
rs12672000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26657882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTGTGCCTTTGTCATGATTTGGTG[G/T]TGTTTTTGTTTGTATGCTGCATACC
Phenotype
MIM: 605215
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SKAP2 PubMed Links

Gene Details

Gene
SKAP2
Gene Name
src kinase associated phosphoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303468.1 1418 Intron NP_001290397.1
NM_003930.4 1418 Intron NP_003921.2
XM_017012771.1 1418 UTR 3 XP_016868260.1

View Full Product Details