Product Details

SNP ID
rs485310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60926343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAACCACAGGCAAATACCTTAAC[C/T]TCTCGGCCTCATTTTCTACATCCAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM109 PubMed Links

Gene Details

Gene
TMEM109
Gene Name
transmembrane protein 109
There are no transcripts associated with this gene.

Gene
TMEM132A
Gene Name
transmembrane protein 132A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017870.3 1030 Intron NP_060340.2
NM_178031.2 1030 Intron NP_821174.1
XM_017017951.1 1030 UTR 5 XP_016873440.1
XM_017017952.1 1030 UTR 5 XP_016873441.1

View Full Product Details