Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001171166.1 | Intron | NP_001164637.1 | |||
NM_001171167.1 | Intron | NP_001164638.1 | |||
NM_001171168.1 | Intron | NP_001164639.1 | |||
NM_015099.3 | Intron | NP_055914.2 | |||
XM_006721478.3 | Intron | XP_006721541.1 | |||
XM_006721481.3 | Intron | XP_006721544.1 | |||
XM_006721482.3 | Intron | XP_006721545.1 | |||
XM_011523746.2 | Intron | XP_011522048.1 | |||
XM_011523747.2 | Intron | XP_011522049.1 | |||
XM_011523748.2 | Intron | XP_011522050.1 | |||
XM_011523749.2 | Intron | XP_011522051.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001167985.1 | Intron | NP_001161457.1 | |||
NM_001167986.1 | Intron | NP_001161458.1 | |||
NM_001167987.1 | Intron | NP_001161459.1 | |||
NM_213726.2 | Intron | NP_998891.2 | |||
XM_005256627.3 | Intron | XP_005256684.1 | |||
XM_005256628.4 | Intron | XP_005256685.1 | |||
XM_005256629.1 | Intron | XP_005256686.1 | |||
XM_006721517.1 | Intron | XP_006721580.1 | |||
XM_006721518.2 | Intron | XP_006721581.1 | |||
XM_006721519.3 | Intron | XP_006721582.1 | |||
XM_011523832.2 | Intron | XP_011522134.1 | |||
XM_011523834.2 | Intron | XP_011522136.1 |