Product Details

SNP ID
rs35281689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7754926 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCTGGAGAGACTGGGGCAGTGA[A/G]GTGGGTAGGGGACCAGGAGCCTGAA
Phenotype
MIM: 608252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFAP1 PubMed Links

Gene Details

Gene
AFAP1
Gene Name
actin filament associated protein 1
There are no transcripts associated with this gene.

Gene
AFAP1-AS1
Gene Name
AFAP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details