Product Details
- SNP ID
-
rs7802779
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:139437833 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAGGAGGGATTGTGTGTTTGAGAT[A/C]ATCTCTGGTCCAGTTCTTTGCCTCT
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLRG2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148579259] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLRG2
- Gene Name
- killer cell lectin like receptor G2
View Full Product Details